Learn More
Applied Biosystems™ ATC Full-skirted Adaptor
Catalog No. A33045
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 adaptor
Catalog No. Quantity
A33045 1 adaptor
1 options
Catalog No. A33045 Supplier Applied Biosystems™ Supplier No. A33045
Add to Cart
Add to Cart
This full-skirted adaptor comes factory-installed on the 96-well Automated Thermal Cycler (ATC).
This full-skirted adaptor comes factory-installed on the 96-well Automated Thermal Cycler (ATC). It ensures that uniform pressure is applied by the heated lid onto the plate, and also ensures that the plate is ejected smoothly without sticking. This adaptor is only for use with 96-well fully skirted plates.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Automated Thermal Cycler (ATC)
Product Type ATC Full-skirted Plate Adaptor
For Use With (Application) End-point PCR
Quantity 1 adaptor
For Research Use Only. Not for use in diagnostic procedures.