Learn More
Applied Biosystems™ ATC Semi-skirted Adaptor
Catalog No. A33044
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Adaptor
Catalog No. Quantity
A33044 1 Adaptor
1 options
Catalog No. A33044 Supplier Applied Biosystems™ Supplier No. A33044
Add to Cart
Add to Cart
This semi-skirted adaptor is for use with the 96-well Automated Thermal Cycler (ATC).
This semi-skirted adaptor is for use with the 96-well Automated Thermal Cycler (ATC). It ensures uniform pressure is applied by the heated lid onto the plate, and also ensures that the plate is ejected smoothly without sticking. This adaptor is only for use with 96-well semi-skirted plates.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Automated Thermal Cycler (ATC)
Product Type ATC Semi-skirted Plate Adaptor
For Use With (Application) End-point PCR
Quantity 1 Adaptor
For Research Use Only. Not for use in diagnostic procedures.