Learn More
ATCC ATCC-HYR0103 hiPSC; Hepatic Fibroblast(r)-2
Supplier: ATCC ACS1007
ATCC-HYR0103 primary hepatic fibroblasts were obtained from a healthy donor. The fibroblasts were reprogrammed by the expression of OCT4, SOX2, KLF4 and MYC gene sequences using retroviral transduction. This cell line provides a unique model system for better understanding hepatic development and differentiation, as well as source material for the development of iPSC-derived hepatocytes. Recommendation: Read the ATCC Stem Cell Culture Guide before initiating your cultures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.