Learn More
ATCC ATCC TC1 Hybridoma Cell Line (CRL-2493), Mouse, IgG1 kappa, Frozen
Supplier: ATCC CRL2493
ATCC TC1 (CRL-2493) is a mouse hybridoma cell line that secretes monoclonal IgG1 kappa antibodies recognizing Pseudomonas aeruginosa exotoxin A. Supplied frozen for research use, the line is suitable for antibody production, immunoassay development, and toxin-related studies.
- Species: Mus musculus.
- Cell type: Hybridoma (B cell-derived).
- Antibody isotype: IgG1 kappa.
- Product format: Frozen vial, stored in liquid nitrogen.
- Biosafety level: 1.
- Applications: Antibody production, ELISA, and immunoblotting.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.