Learn More
ATCC ATCC VR-1607 Human rhinovirus 37, strain 151-1 - frozen viral isolate for research use only
Supplier:  ATCC VR1607
Encompass
Human rhinovirus 37 strain 151-1 is a mycoplasma-free, fully sequenced viral isolate supplied as a frozen preparation for laboratory research in respiratory and infectious disease studies. It is intended for research use only and not for diagnostic or therapeutic use.
  • Frozen preparation; store at -70°C or colder.
  • Genome verified by whole-genome sequencing.
  • Mycoplasma not detected in this preparation.
  • Propagated in H1HeLa cells; alternate human cell lines reported.
  • Produces cytopathic effects such as cell rounding and sloughing.
  • Intended for laboratory research use only.
Catalog No. NC3834026