Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG12-ATG5 Recombinant Rabbit Monoclonal Antibody (20H20L20)

Catalog No. 702433 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-243-3 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-243-3 Supplier Invitrogen™ Supplier No. 702433
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Pig, Cat and Mouse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG12, another member of the autophagy protein family, forms a conjugate with ATG5; this conjugate has a ubiquitin-protein ligase (E3)-like activity for protein lipidation in autophagy. This conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. ATG12 has also been shown to interact with ATG10 in human embryonic kidney cells in the presence of ATG7. At least two isoforms of ATG12 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG12-ATG5
Applications Western Blot
Classification Recombinant Monoclonal
Clone 20H20L20
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene ATG12
Gene Accession No. O94817, Q2TBJ5, Q3MQ06, Q99J83, Q9CQY1, Q9H1Y0
Gene Alias 2010107M05Rik; 3110067M24Rik; 4931423H11Rik; A330058M13Rik; Apg12; Apg12 (autophagy, yeast) homolog; APG12L; APG12-like; APG5; Apg5l; APG5-LIKE; apoptosis-specific protein; ASP; Atg12; ATG12 autophagy related 12 homolog; Atg12l; Atg5; ATG5 autophagy related 5 homolog; Atg5l; Autophagy protein 5; autophagy related 12; autophagy related 5; autophagy-related 12; autophagy-related 5; autophagy-related protein 12; AW319544; C88337; FBR93; HAPG12; hAPG5; Paddy; Ubiquitin-like protein ATG12
Gene Symbols ATG12, ATG5
Host Species Rabbit
Immunogen Peptides corresponding to Human ATG12 (aa 43-μ, 122-133).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11793, 361321, 365601, 67526, 9140, 9474
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.