Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG16L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106824 Supplier Invitrogen™ Supplier No. PA5106824
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATG16L2.

May play a role in autophagy. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG16L2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATG16L2
Gene Accession No. Q6KAU8, Q8NAA4
Gene Alias 2410118P20Rik; APG16-like 2; ATG16 autophagy related 16-like 2; ATG16B; Atg16l2; Atg16L2 alpha; Atg16L2 beta; autophagy related 16 like 2; autophagy related 16-like 2; autophagy related 16-like 2 (S. cerevisiae); autophagy-related 16-like 2 alpha; autophagy-related 16-like 2 beta; autophagy-related protein 16-2; mFLJ00012 protein; RGD1311400; WD repeat-containing protein 80; WDR80
Gene Symbols ATG16L2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATG16L2(Accession Q8NAA4), corresponding to amino acid residues L242-K292.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308865, 73683, 89849
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.