Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Atherogenic Diet

Semi-purified atherogenic diet modified for mice.

$898.00 - $3295.00

Specifications

Product Type Animal Diet
For Use With (Application) Rodent diet
Storage Requirements 2°C to 8°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP296040410
View Documents
MP Biomedicals™
0296040410
10 kg
Each for $898.00
Only null left
Add to Cart
 
MP296040420
View Documents
MP Biomedicals™
0296040420
20 kg
Each for $1,533.00
Only null left
Add to Cart
 
MP296040450
View Documents
MP Biomedicals™
0296040450
50 kg
Each for $3,295.00
Only null left
Add to Cart
 
Description

Description

  • Atherogenic diet is rich in cholesterol and other atherogenic factors
  • Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease
Specifications

Specifications

Animal Diet
Rodent diet
2°C to 8°C
Videos
SDS
Documents

Documents

Product Certifications

Atherogenic diet is rich in cholesterol and other atherogenic factors. Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.