Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ATI 2341
GREENER_CHOICESDP

Catalog No. 57951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57951 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 57951 Supplier Tocris Bioscience™ Supplier No. 5795/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

CXCR4 allosteric agonist

ATI 2341 is a CXCR4 allosteric agonist (EC50 = 194 nM). Induces chemotaxis in CXCR4-expressing cells in vitro. Mobilizes polymorphonuclear neutrophils (PMNs) and hematopoietic stem and progenitor cells (HSPCs) from bone marrow niche in vivo. Exhibits biased signaling towards Gi proteins over G13 and β-arrestin.

Specifications

Purity 0.95
CAS 1337878-62-2
Content And Storage Store at -20°C
Description Chemokine Receptor Agonists
Molecular Weight (g/mol) Observed M.W.: 2256.82
Quantity 1 mg
Structural Form C104H178N26O25S2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.