Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATL1 Mouse anti-Human, Clone:1F6B12, Abnova™

Catalog No. 89334404 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-404 Supplier Abnova Corporation Supplier No. MAB17914
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human ATL1.

The protein encoded by this gene is a GTPase and a Golgi body transmembrane protein. The encoded protein can form a homotetramer and has been shown to interact with spastin and with mitogen-activated protein kinase kinase kinase kinase 4. This protein may be involved in axonal maintenance as evidenced by the fact that defects in this gene are a cause of spastic paraplegia type 3. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen ATL1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F6B12
Conjugate Unconjugated
Description atlastin GTPase 1
Dilution ELISA (1:10000)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ATL1
Gene Alias AD-FSP, FSP1, GBP3, SPG3, SPG3A, atlastin1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-100 of human ATL1.
Quantity 100 μg
Gene ID (Entrez) 51062
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.