Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106923 Supplier Invitrogen™ Supplier No. PA5106923
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATL1.

The protein encoded by this gene is a GTPase and a Golgi body transmembrane protein. The encoded protein can form a homotetramer and has been shown to interact with spastin and with mitogen-activated protein kinase kinase kinase kinase 4. This protein may be involved in axonal maintenance as evidenced by the fact that defects in this gene are a cause of spastic paraplegia type 3. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATL1
Gene Accession No. Q6PST4, Q8BH66, Q8WXF7
Gene Alias 4930435M24Rik; Adfsp; AD-FSP; ATL1; ATL1 alpha; ATL1 beta; ATL1 gamma; ATL1-delta; atlastin GTPase 1; atlastin1; atlastin-1; Bcl11B; BCL-11B; BCL11B/TRDC fusion; brain-specific GTP-binding protein; Ctip 2; CTIP2; CTIP-2; Fsp1; GBP3; GBP-3; GTP-binding protein 3; guanine nucleotide-binding protein 3; guanylate-binding protein 3; hGBP3; hRit1; hRIT1 alpha; HSN1D; Rit 1; Rit1; spastic paraplegia 3 protein A; Spastic paraplegia 3A homolog; SPG3; SPG3A; ZNF856B
Gene Symbols ATL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATL1(Accession Q8WXF7), corresponding to amino acid residues K78-E128.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362750, 51062, 73991
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.