Learn More
SHOWA™ KV300 Aramid Fiber Cut-Resistant Gloves, Natural Rubber Coating
Shop All Showa Best Glove ProductsDescription
- Aramid fiber gloves provide ANSI Level A3 Cut Resistance
- Rough-textured latex coating enhances wet and dry grip
- Uncoated knuckles ensure flexibility and precision
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Construction | Seamless Knit |
| Coating Material | Latex |
| Color | Yellow, Blue |
| Cut Resistance Rating | ANSI A3 |
| Exterior Finish | Fully Textured |
| Hand | Specific |
| Material | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Small |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
