Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ KV300 Aramid Fiber Cut-Resistant Gloves, Natural Rubber Coating

Catalog No. 19270237 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-270-237 X-Large
19-270-234 Small
19-270-235 Medium
19-270-236 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-270-237 Supplier SHOWA™ Supplier No. KV30010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves feature a 10-gauge, seamless knit liner with a latex coating for enhanced grip capabilities in wet or dry conditions.

  • Aramid fiber gloves provide ANSI Level A3 Cut Resistance
  • Rough-textured latex coating enhances wet and dry grip
  • Uncoated knuckles ensure flexibility and precision

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Latex
Color Yellow, Blue
Cut Resistance Rating ANSI A3
Exterior Finish Fully Textured
Hand Specific
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:3X43CP
For Use With (Application) Heavy equipment, bottling, glass and windows, bottling and canning, police and defense purposes
Glove Cuff Style Knit
Model KV300-10
Puncture Rating ANSI/ISEA 105-16 Level 5
Series KV300
Yarn Gauge 10
Length (English) 9.5 in.
Length (Metric) 24.1 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.