Learn More
ECM Bioscience ATM Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience AK6300
Encompass_Preferred
The ATM phospho-regulation antibody sampler kit can be used to detect ATM phosphorylation on Ser-794 and Ser-1981. The kit also includes a monoclonal antibody to monitor total ATM expression levels and secondary reagents for detection of these antibodies. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8598
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.