Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ AtmoSense Oxygen Monitor
SDP

Catalog No. 16985666 Shop All Labconco Corporation Products
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
16-985-666 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 16-985-666 Supplier Labconco™ Supplier No. 5255590
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

For use with Labconco Glove Boxes for general purpose trace oxygen detection as low as 1 ppm.

  • Digital LCD readout from 0 to 1000 ppm oxygen with resolution accuracy of 0.1 ppm from 0 to 200 ppm and a resolution accuracy of 1 ppm from 200 to 1000 ppm.
  • 3 user-configurable O2 alarm levels and relays.

Specifications

Product Type Oxygen Monitor
Voltage 115 V
For Use With (Application) Low-range Oxygen Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.