Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Atoms, Electrons And Energy Kit
SDPSCI_ED

Catalog No. S00663 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S00663 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00663 Supplier United Scientific™ Supplier No. AEEKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Atoms, Electrons and Energy Kit takes a tradtionally difficult concept and enables students to learn hands on.

  • This kit contains materials representing atomic structures and electron arrangement within elements.
  • It also provides materials necessary to demonstrate how atomic arrangement relates to placement in the periodic table of elements.
  • The kit contains an energy level template, and enough nuclear areas, orbitals (s,p,d) and electrons to represent the first 36 elements.
  • Each nucleus and orbital are color coded to match a large periodic chart depicting trends in electron orbital arrangement and electronegativity.
  • Also included is a large 1st ionization energy chart for these 36 elements.
  • Included are 10 sets of smaller, reusable student manipulatives suitable for use by up to 30 students.
  • Link and password provided to downloadable digital activity guide and teacher resources.

Specifications

Grade 6 to 12
Includes 1st Ionization energy chart
Class Size 30 Students
Product Type STEM Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.