Learn More
Cayman Chemical Atorvastatin (calcium salt), 134523-03-8, 5 mg
SDP
Supplier:  Cayman Chemical 104935
Encompass_Preferred
Molecular Formula C33H35FN2O5 • 1/2Ca, Purity ≥98%, Formula Weight 578.7, Synonyms: 2-(4-fluorophenyl)-βR,δR-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid, hemicalcium salt; An HMG-CoA reductase inhibitor (IC50s = 73, 102, and 0.6 nM for HepG2 cells, human fibroblasts, and rat hepatocytes, respectively).
Catalog No. NC0754538
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.