Learn More
Medchemexpress LLC HY-B2176A 100mg Medchemexpress, ATP (disodium trihydrate) CAS:51963-61-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2176A100MG
Encompass_Preferred
Medchemexpress, HY-B2176A 100mg ATP (disodium trihydrate) CAS:51963-61-2 ATP disodium trihydrate (Adenosine 5'-triphosphate disodium trihydrate) is a central component of energy storage and metabolism in vivo. ATP disodium trihydrate provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP disodium trihydrate is an important endogenous signaling molecule in immunity and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2731
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.