Learn More
Enzo Life Sciences Adenosine 5'-O-(3-thiotriphosphate) . tetralithium salt (5mg). CAS: 93839-89-5
SDP
Supplier:  Enzo Life Sciences ALX480066M005
Encompass_Preferred
ATP analog that can substitute ATP in various kinase reactions. Substrate and inhibitor of ATP-dependent enzyme systems. The resulting thiophosphorylated proteins are resistant to dephosphorylation by protein phosphatases. Increases the activity of Ca2 -activated K channels. Alternative name: ATP-gamma-S . 4Li. Purity: ≥90% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9315
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.