Learn More
Abcam ATP Synthase, Relative Activity Microplate Assay Kit, 96 Tests
Description
- Assay Type: Enzyme activity
- Detection Method: Colorimetric
- Sample Types: Cell lysate, cell culture extracts, cell culture supernatant, tissue, tissue extracts
- Reactive Species: Rat, cow, human
- Applications: Enzyme activity assay
- Features: Measures ATP synthase activity in cell/tissue extracts and cell culture media; compatible with microplate readers; cited in over 16 publications.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | ATP Synthase |
| Reactivity | Human, Cow, Rat |
| No. of Reactions | 96 Tests |
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Microplate Reader |
| Detection Method | Colorimetric |
| Format | Microplate Assay Kit |
| Cross Reactivity | Rat, Cow, Human |
| Sample Type | Cell Lysate, Cell Culture Extracts, Cell Culture Supernatant, Tissue, Tissue Extracts |
| Quantity | 96 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.