Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP2c1/2 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501984725
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4725 Supplier Bioss Supplier No. BS2434R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of calcium.

Specifications

Antigen ATP2c1/2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATP2C1
Gene Accession No. A7L9Z8, O75185, P98194, Q64566, Q80XR2, Q8R4C1
Gene Symbols ATP2C1, ATP2C2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATP2c1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170699, 171496, 235574, 27032, 69047, 9914
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.