Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP synthase Monoclonal Antibody (12F4AD8AF8)
GREENER_CHOICE

Catalog No. 439700
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-970-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-970-0 Supplier Invitrogen™ Supplier No. 439700
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Bovine heart mitochondria.

Complex V, also called F1F0ATPase or ATP synthase, is responsible for ATP production in oxidative phosphorylation and can work in reverse as a proton pumping ATPase. The enzyme was thought to be localized exclusively to mitochondria. However, it has recently been identified on the plasma membrane of several cell types including hepatocytes where it functions as the HDL receptor, on endothelial cells where it may act as the angiostatin receptor, and on the surface of cancer cells. The enzyme in mammals is composed of 17 subunits, five of which make up the easily detached F1. The remainder subunits are components of two stalk domains and the proton pumping F0 part of the machinery. Two of the subunits of the F0 part are encoded on mitochondrial DNA while the other subunits are nuclear encoded. Mutations in the mitochondrial-encoded subunits of ATP synthase (Complex V) cause OXPHOS disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP synthase
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 12F4AD8AF8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene ATP5PD
Gene Accession No. O75947, O75964, P00829, P00846, P00848, P03928, P03930, P05496, P05632, P06576, P13620, P18859, P19483, P24539, P25705, P30049, P36542, P48047, P48201, P56134, P56135, P56378, P56381, P56382, P56385, P56480, P97450, Q06055, Q06185, Q91VR2, Q9CPQ8, Q9CQQ7, Q9CR84, Q9D3D9, Q9DB20, Q9DCX2, Q9UII2
Gene Alias 0610008F14Rik; 0610009D10Rik; 1110019H14Rik; 1500000I11Rik; 1700094F02Rik; 1810041M08Rik; 2410043G19Rik; 2610008D24Rik; 4933437C06Rik; 6.8 kDa mitochondrial proteolipid; 6.8 kDa mitochondrial proteolipid protein; A6L; AA960090; AI876556; alpha subunit ATP synthase isoform precursor (EC 3.6.1.34); ATP synthase alpha chain, mitochondrial; ATP synthase alpha subunit; ATP synthase alpha subunit precursor; ATP synthase B chain, mitochondrial; ATP synthase c subunit; ATP synthase D chain, mitochondrial; ATP synthase e chain, mitochondrial; ATP synthase epsilon chain, mitochondrial; ATP synthase f chain, mitochondrial; ATP synthase F(0) complex subunit B1, mitochondrial; ATP synthase F(0) complex subunit C1, mitochondrial; ATP synthase F(0) complex subunit C2, mitochondrial; ATP synthase F(0) complex subunit C3, mitochondrial; ATP synthase F0 subunit 6; ATP synthase F0 subunit 8; ATP synthase F1 subunit alpha; ATP synthase F1 subunit beta; ATP synthase F1 subunit delta; ATP synthase F1 subunit epsilon; ATP synthase F1 subunit gamma; ATP synthase g chain, mitochondrial; ATP synthase gamma chain, mitochondrial; ATP synthase gamma-subunit gene encoding a mitochondrial protein; ATP synthase inhibitor protein; ATP synthase lipid-binding protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit 6.8PL; ATP synthase membrane subunit c locus 1; ATP synthase membrane subunit c locus 2; ATP synthase membrane subunit c locus 3; ATP synthase membrane subunit e; ATP synthase membrane subunit f; ATP synthase membrane subunit g; ATP synthase peripheral stalk subunit d; ATP synthase peripheral stalk subunit F6; ATP synthase peripheral stalk subunit OSCP; ATP synthase peripheral stalk-membrane subunit b; ATP synthase protein 8; ATP synthase proteolipid P1; ATP synthase proteolipid P2; ATP synthase proteolipid P3; ATP synthase proton-transporting mitochondrial F(0) complex subunit B1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C2; ATP synthase proton-transporting mitochondrial F(0) complex subunit C3; ATP synthase subunit 9; ATP synthase subunit a; ATP synthase subunit alpha, mitochondrial; ATP synthase subunit ATP5MPL, mitochondrial; ATP synthase subunit b; ATP synthase subunit b, mitochondrial; ATP synthase subunit beta, mitochondrial; ATP synthase subunit d, mitochondrial; ATP synthase subunit delta, mitochondrial; ATP synthase subunit e, mitochondrial; ATP synthase subunit e, mitochondrial, N-terminally processed; ATP synthase subunit epsilon, mitochondrial; ATP synthase subunit f, mitochondrial; ATP synthase subunit g, mitochondrial; ATP synthase subunit gamma, mitochondrial; ATP synthase subunit O, mitochondrial; ATP synthase, H+ transporting mitochondrial F1 complex, alpha subunit; ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C3 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d isoform a; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit E; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit f, isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F6
Gene Symbols ATP5F1A, ATP5F1B, ATP5F1C, ATP5F1D, ATP5F1E, ATP5IF1, ATP5MC1, ATP5MC2, ATP5MC3, ATP5ME, ATP5MF, ATP5MG, ATP5MPL, ATP5PB, ATP5PD, ATP5PF, ATP5PO, MT-ATP6, MT-ATP8
Host Species Mouse
Immunogen Bovine Complex V.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10476, 10632, 11947, 11949, 11950, 11951, 11957, 11958, 17705, 17706, 27425, 28080, 282578, 282710, 327675, 4508, 4509, 498, 506, 509, 513, 514, 515, 516, 517, 518, 521, 522, 539, 57423, 617230, 66043, 67126, 71679, 93974, 9551, 9556
Target Species Human, Mouse, Bovine
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.