Learn More
Invitrogen™ ATP5I Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 8 kDa. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | ATP5I |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Atp5me |
| Gene Accession No. | P56385, Q06185 |
| Gene Alias | 2610008D24Rik; ATP synthase e chain, mitochondrial; ATP synthase membrane subunit e; ATP synthase subunit e, mitochondrial; ATP synthase subunit e, mitochondrial, N-terminally processed; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit E; ATP synthase, H+ transporting, mitochondrial F1F0 complex, subunit e; ATP synthase, H+ transporting, mitochondrial Fo complex subunit E; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit E; Atp5i; Atp5k; Atp5me; ATPase subunit e; F1F0-ATP synthase, murine e subunit; Lfm1; Lfm-1 |
| Gene Symbols | Atp5me |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.