Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5J2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575692 Supplier Invitrogen™ Supplier No. PA575692
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ATP5J2 is part of the complex F0 domain of the mitochondrial ATP synthase. This produces ATP from ADP in the presence of a proton gradient across the membrane which is generated by electron transport complexes of the respiratory chain. It is composed of two complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, which comprises the proton channel. ATP5J2 encodes the f subunit of the Fo complex. There are 2 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5J2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP5MF
Gene Accession No. D3ZAF6, P56134, P56135
Gene Alias 1110019H14Rik; ATP synthase f chain, mitochondrial; ATP synthase membrane subunit f; ATP synthase subunit f, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit f, isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F2; ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F2; ATP5J2; ATP5JL; Atp5mf; F1F0-type ATPase subunit f; F1Fo-ATP synthase complex Fo membrane domain f subunit; F1Fo-ATPase synthase f subunit; hypothetical protein, clone:2-31
Gene Symbols ATP5MF
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human ATP5J2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57423, 690441, 9551
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.