Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1G3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501732434 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2434 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2434 Supplier Proteintech Group Inc Supplier No. 195231AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

ATPase, H+ transporting, lysosomal (vacuolar proton pump) subunit G3,ATPase, H+ transporting, lysosomal 13kD, V1 subunit G,ATPase, H+ transporting, lysosomal, V1 subunit G3,OTTHUMP00000033686,V-ATPase 13 kDa subunit 3,V-ATPase G subunit 3,V-ATPase G3 subunit

Specifications

Antigen ATP6V1G3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP6V1G3
Gene Accession No. Q8BMC1, Q96LB4
Gene Alias ATP6G3, ATP6V1G3, V ATPase 13 kDa subunit 3, V ATPase subunit G 3, Vma10
Gene Symbols Atp6v1g3
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127124, 338375
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.