Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP8 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735435 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5435 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5435 Supplier Proteintech Group Inc Supplier No. 267231AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mitochondrial ATP synthase is composed of two multi-subunit complexes that utilize an inner membrane electrochemical gradient to catalyze the synthesis of ATP during oxidative phosphorylation. The two multi-subunit complexes are designated F1 and F0, the former of which comprises the soluble catalytic core and the latter of which comprises the membrane-spanning proton channel of ATP synthase. ATP8, also known as MT-ATP8, ATPASE8 or MTATP8, is a 68 amino acid single-pass membrane protein that localizes to mitochondria and functions as a minor subunit of the F0 complex. The gene encoding ATP8 maps to the mitochondrial chromosome and may be involved in the pathogenesis of multiple sclerosis.

Specifications

Antigen ATP8
Applications Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ATP8
Gene Accession No. P03928
Gene Alias A6L, ATP synthase protein 8, ATP8, ATPASE8, F ATPase subunit 8, MT ATP8, MTATP8
Gene Symbols ATP8
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4509
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.