Learn More
Abcam Atpenin A5, Mitochondrial Complex II Inhibitor, Solid, 1 mg
Description
- Molecular weight: 366.2 Da
- Purity: >95%
- Form: Solid
- Activity: Highly selective, potent ubiquinone-binding site mitochondrial complex II inhibitor (IC50 values are 12 and 3.7 nM for nematode and mammalian mitochondria respectively)
- Key biological effects: Stimulates mitochondrial KATP channels; shows anti-ischemic and cardioprotective effects in vivo
- Chemical composition: 3-[(2S,4S,5R)-5,6-dichloro-2,4-dimethylhexanoyl]-4-hydroxy-5,6-dimethoxy-1H-pyridin-2-one
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Mitochondrial Complex II |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 1 mg |
| For Use With (Application) | Mitochondrial complex II inhibition, KATP channel stimulation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.