Learn More
Crescent Chemical Co Inc ATRAZINE-2-HYDROXY SOLN 1 ML
SDP
Supplier:  Crescent Chemical Co Inc PSXA10333000ME
Encompass_Preferred
Atrazine-2-hydroxy 100 µg/mL in Methanol Herbicide (Metabolite solution) CAS 2163-68-0 Molecular Weight - 197.2376 Storage - RT Catalog Num - DRE-XA10333000ME chemical
Catalog No. 50-798-67
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.