Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atrazine 97.0+%, TCI America™
SDP

Catalog No. A165025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A165025G 25 g
A16505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A165025G Supplier TCI America Supplier No. A165025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1912-24-9
Molecular Formula C8H14ClN5
Molecular Weight (g/mol) 215.69
MDL Number MFCD00041810
InChI Key MXWJVTOOROXGIU-UHFFFAOYSA-N
Synonym atrazine, gesaprim, chromozin, oleogesaprim, aktikon, argezin, atazinax, atranex, atrasine, atrazin
PubChem CID 2256
ChEBI CHEBI:15930
IUPAC Name 6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
SMILES CCNC1=NC(Cl)=NC(NC(C)C)=N1

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 215.69
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Atrazine
EINECSNumber (5)-3851
RTECSNumber XY5600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.