Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Atrial Natriuretic Peptide (11-30), Frog

Catalog No. AAJ66748EXD
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66748EXD 0.5 mg
AAJ66748MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66748EXD Supplier Thermo Scientific™ Supplier No. J66748EXD

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Species Frog
Purification Method HPLC
Common Name ANP (11-30), frog
Content And Storage -30°C to -10°C
Format Lyophilized Powder
Molecular Weight (g/mol) 2116.40
Product Type Atrial Natriuretic Peptide (11-30), frog
Quantity 0.5 mg
Regulatory Status RUO - research use only
Sequence Cys-Phe-Gly-Ser-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Met-Gly-Cys-Gly-Arg-Phe (Disulfide bridge Cys11-Cys27)

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.