Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATRNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590091 Supplier Invitrogen™ Supplier No. PA590091
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYAQVSQSKP CERTGSCFSG RCVNSTCLCD PGWVGDQCQH CQGRFKLTEP SGYLTDGPIN YKYKTKCTWL IEGYPNAVLR LRFNHFATEC SWDHMYVYDG DSIYAPLI; Positive Samples: Mouse liver; Cellular Location: Membrane, Single-pass type I membrane protein.

ATRNL1 (Attractin Like 1) is a Protein Coding gene. May play a role in melanocortin signaling pathways that regulate energy homeostasis. Gene Ontology (GO) annotations related to this gene include carbohydrate binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATRNL1
Applications Western Blot
Classification Polyclonal
Concentration 2.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ATRNL1
Gene Accession No. Q5VV63, Q6A051
Gene Alias AI504415; Alp; Atrnl; Atrnl1; Atrnlp; attractin like 1; attractin-like 1; attractin-like protein 1; AW555641; bA338L11.1; bA454H24.1; Kiaa0534; RGD1563485
Gene Symbols ATRNL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 53-160 of human ATRNL1 (NP_0012632111).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226255, 26033
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.