Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atrolactic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. A10351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10351G 1 g
A10355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10351G Supplier TCI America Supplier No. A10351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 515-30-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00004451
InChI Key NWCHELUCVWSRRS-UHFFFAOYSA-N
Synonym atrolactic acid, 2-phenyllactic acid, 2-hydroxy-2-phenylpropionic acid, alpha-methylmandelic acid, phenyllactic acid, 2-phenyl-2-hydroxypropionic acid, dl-atrolactic acid, alpha-hydroxy-alpha-phenylpropionic acid, mandelic acid, .alpha.-methyl, benzeneacetic acid, .alpha.-hydroxy-.alpha.-methyl
PubChem CID 1303
ChEBI CHEBI:50392
IUPAC Name 2-hydroxy-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)(C(=O)O)O

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Atrolactic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.