Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Atrophin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556799
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556799 Supplier Invitrogen™ Supplier No. PA556799
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HPYAMSPSLG SLRPYPPGPA HLPPPHSQVS YSQAGPNGPP VSSSSNSSSS TSQGSYPCSH PSPSQGPQGA PYPFPPVPTV TTSSATLSTV IATVASSPAG YKTASPPGPP PYGKRAPSPG AYKTATPPGY KPGSPPSFRT GTPPGY Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Dentatorubral pallidoluysian atrophy (DRPLA) is a rare neurodegenerative disorder characterized by cerebellar ataxia, myoclonic epilepsy, choreoathetosis, and dementia. The disorder is related to the expansion from 7-35 copies to 49-93 copies of a trinucleotide repeat (CAG/CAA) within this gene. The encoded protein includes a serine repeat and a region of alternating acidic and basic amino acids, as well as the variable glutamine repeat. Alternative splicing results in two transcripts variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atrophin 1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATN1
Gene Accession No. P54259
Gene Alias ATN1; Atr1; atrophin 1; Atrophin1; atrophin-1; B37; D12S755E; dentatorubral pallidoluysian atrophy; dentatorubral-pallidoluysian atrophy protein; Dentatorubral-pallidoluysian atrophy protein homolog; Drpla; HRS; NOD
Gene Symbols ATN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Atrophin 1. Recombinant protein control fragment (Product #RP-91651).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.