Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atropine Sulfate Monohydrate 99.0+%, TCI America™
SDP

Catalog No. A055025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A055025G 25 g
A05505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A055025G Supplier TCI America Supplier No. A055025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5908-99-6
Molecular Formula C17H27NO8S
Molecular Weight (g/mol) 405.462
MDL Number MFCD00077268
InChI Key PVGPXGKNDGTPTD-IJTOKZDFSA-N
Synonym atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid
PubChem CID 23624044
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O

Specifications

Melting Point 194°C
Color White
Quantity 25 g
UN Number 1544
Formula Weight 676.82
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Atropine Sulfate Monohydrate
EINECSNumber (5)-3898
RTECSNumber CK2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.