Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atropine 99.0+%, TCI America™
SDP

Catalog No. A075425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A075425G 25 g
A07545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A075425G Supplier TCI America Supplier No. A075425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51-55-8
Molecular Formula C17H23NO3
Molecular Weight (g/mol) 289.375
MDL Number MFCD00022622
InChI Key RKUNBYITZUJHSG-PJPHBNEVSA-N
Synonym r-atropine, +-atropine, +-hyoscyamine, hyoscyamine, tropan-3alpha-yl 2r-3-hydroxy-2-phenylpropanoate, 3-endo-8-methyl-8-azabicyclo 3.2.1 oct-3-yl 2r-3-hydroxy-2-phenylpropanoate, atropine sulfate, unii-v3203si72m, 1s,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2r-3-hydroxy-2-phenylpropanoate, prestwick0_000233
PubChem CID 174174
IUPAC Name [(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3

Specifications

Melting Point 116°C
Color White
pH 10.0 (0.0015 mol H2O soln.)
Quantity 25 g
UN Number 1544
Formula Weight 289.38
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Atropine
RTECSNumber CK0700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.