Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATRX Mouse anti-Mouse, Human, Clone: 39F, MilliporeSigma™

Catalog No. MABE1798MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1798MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1798MI Supplier MilliporeSigma Supplier No. MABE1798
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ATRX Antibody, clone 39f is a highly specific mouse monoclonal antibody that targets ATRX and has been tested in Immunocytochemistry and Western Blotting .
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATRX
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 39F
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ATRX,RAD54L,XH2
Host Species Mouse
Immunogen Recombinant fragment corresponding to 235 amino acids from the N-terminal half of human ARTX.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000480
Test Specificity Clone 39f specifically recognizes human ARTX. This clone binds to an epitope beetween amino acid 85-319 in the N-terminal half of human ATRX.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.