Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega AttoPhos™ Buffer

Catalog No. PRS1022 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Substrate Quantity:
60mL
240mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Substrate Quantity
PRS1022 240mL
PRS1021 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRS1022 Supplier Promega Supplier No. S1022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

AttoPhos Substrate, AttoPhos Buffer, AttoPhos Calibration Solution (Mfr. No. S1000 only)

The AttoPhos(R) Buffer is optimized for sensitive detection of alkaline phosphatase (AP) when used to prepare working solutions with AttoPhos(R) Substrate.

  • Produces a low fluorescence signal until enzymatically acted upon, yielding detection of AP to 0.1 attomole
  • Provides low background
  • Offers linear kinetics over five orders of magnitude of AP concentration
  • Has an unusually large Stokes' shift (≈120nm), which leads to higher detection sensitivity

Specifications

Includes AttoPhos buffer
pH 10
Storage Requirements 4°C
Substrate Quantity 240mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.