Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Attune™ Focusing Fluid (1X)

Catalog No. 501034959
Encompass
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-103-4959 6 x 1 L
44-497-91 6 x 1 L
A24904 10 L
44-886-21 1 L
J106627 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-103-4959 Supplier THERMO SCI LSG 3P Supplier No. 4449791

Attune™ Focusing Fluid (1X)

Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.

Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.

Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.

High Quality
Attune™ Focusing Fluid is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Product Type Focusing Fluid
Content And Storage Store at 15—30°C
For Use With (Equipment) Attune™ Acoustic Focusing Cytometer
Format Liquid
Quantity 6 x 1 L
What is the smallest size that I can detect with the Attune NxT Acoustic Focusing Cytometer?

The smallest size that you can detect with the Attune NxT Acoustic Focusing Cytometer is 0.5 µm.

What is the Attune NxT Autosampler?

The Attune NxT Autosampler, an optional accessory for the Attune NxT Acoustic Focusing Cytometer, enables rapid processing of up to 384 samples. It has broad compatibility with different plate formats, both 96- and 384-well plates. It has an intelligent probe designed to minimize clogging and carryover (<0.5%) and to prevent damage to the instrument. It mixes by aspiration rather than shaking to ensure homogeneity of the sample and maintain cell viability. Is performs automated cleaning as part of the shutdown process of the Attune NxT Cytometer. It provides consistent data regardless of sampling method (tube vs. plate) and collection rate.

What are the advantages of acoustic-assisted hydrodynamic focusing in flow cytometry?

-Modular design - Multiple configurations available - field upgradable.
-Save time - 10X faster speeds with no loss in data quality.
-Simplified sample prep - No wash, no lyse options, non-clogging fluidics.
-Enables unique applications - Complex protocols on a broad range of cell types and samples.

How is the Attune NxT Acoustic Focusing Cytometer different from traditional flow cytometers?

With the option to be configured with up to 4 lasers and 14 colors for multi-parameter analysis the Attune NxT Acoustic Focusing Cytometer was designed as a modular system to fit most experimental needs and lab budgets. The novel design of the optical path helps ensure precise fixed alignment of four spatially separated lasers onto the sample stream enabling consistency in data over time, superior performance, and superior reliability. The instrument can be configured with up to 4 solid-state lasers (405 nm, 488 nm, 561 nm, and 637 nm) with flat top beam profiles.

The Attune NxT Flow Cytometer's acoustic focusing uses ultrasonic radiation pressure (> 2 MHz) to transport particles into the center of the sample stream. This pre-focused stream is then injected into the sheath stream, which supplies an additional hydrodynamic pressure to the sample. The combination of these two forces- termed acoustic-assisted hydrodynamic focusing-results in a narrow core stream and uniform laser illumination, regardless of the sample input rate. In traditional cytometers that rely solely on hydrodynamic focusing, the sample core widens to accommodate the increases in flow rate, which results in less uniform laser light illumination.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.