Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATXN3L Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734009 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4009 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4009 Supplier Proteintech Group Inc Supplier No. 231661AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

ATXN3L, also named as Ataxin-3-like protein, is a 355 amino acid protein, which contains 1 Josephin domain and 2 UIM (ubiquitin-interacting motif) domains. ATXN3L localizes in the nucleus and as a deubiquitinating enzyme that cleaves both ′Lys-48′-linked and ′Lys-63′-linked poly-ubiquitin chains.

Specifications

Antigen ATXN3L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATXN3L
Gene Accession No. Q9H3M9
Gene Alias ataxin 3 like, ATX3L, ATXN3L, MJDL, Putative ataxin 3 like protein
Gene Symbols ATXN3L
Host Species Rabbit
Immunogen ATXN3L Fusion Protein Ag19611
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92552
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.