Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATXN7L2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988175
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8175 Supplier Bioss Supplier No. BS7976R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCA7 is an autosomal dominant neurodegenerative disorder characterized by ataxia and selective neuronal cell loss caused by the expansion of a translated CAG repeat encoding a polyglutamine tract in ataxin-7, which is the SCA7 gene product. Ataxin-7 is a nuclear protein that is expressed within neurons both affected and unaffected in SCA7 pathology with subcellular localization being variable depending upon the neuronal subtype. Polyglutamine expanded in ataxin-7 may carry out its pathogenic effects in the nucleus by altering the matrix-associated nuclear structure and/or by disrupting nucleolar function.ATXN7L2 (Ataxin-7-like protein 2) is a 722 amino acid protein that contains a SCA7 domain, which is highly conserved through all members of the ATXN7 gene family. The gene encoding ATXN7L2 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome.

Specifications

Antigen ATXN7L2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATXN7L2
Gene Accession No. Q5T6C5
Gene Alias 2610528J18Rik; ataxin 7 like 2; ataxin 7-like 1; ataxin 7-like 2; Ataxin-7-like protein 2; ATXN7L2; mFLJ00381 protein; RGD1307047
Gene Symbols ATXN7L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATXN7L2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127002
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.