Learn More
United Scientific™ Auditory Conductivity Indicator
SDPSCI_ED
Catalog No. S000871 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Alarm Output:
Buzzer/Light
Catalog No. Alarm Output
S000871 Buzzer/Light
1 options
Prop 65
Catalog No. S000871 Supplier United Scientific™ Supplier No. AUDCND
Add to Cart
Add to Cart
Prop 65
This Indicator is an excellent alternative to light only indicators.
Investigate the conductivity of a solution with this excellent alternative to light only indicators. The current flow to a buzzer changes with the conductivity of a solution. The more free ions in a solution, the better it conducts electricity and the louder the buzzer sounds. Includes an on/off button with an LED to indicate if the unit is turned on. Includes activity guide. Requires a 9 V battery (not included).
  • Buzzer provides an audible indicatorof the conductivity of a solution
  • Reusable design
  • Includes activity guide
Specifications
Measurement Modes Conductivity
Range (Conductivity) Yes or No
Range (Salinity) None
Range (TDS) None
Temperature Range (Metric) None
Range (Resistivity) None
Display Type None
Data Points Single Set Point
Alarm Output Buzzer/Light
IP Rating None
Material Plastic
Type Conductivity Indicator