Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
AUNP 12 is an immune checkpoint modulator targeting the programmed cell death 1 (PD-1) signaling pathway. It inhibits the interaction between PD-1 and its ligands, PD-L1 and PD-L2. In the mouse splenocyte assay system, AUNP 12 rescues proliferation (EC50 values are 17 and 16 nM for PD-L1 and PD-L2, respectively) and enhances T-cell activity by restoring IFNγ release inhibited by PD-L1 and PD-L2 (EC50 values are 49 nM and 51 nM respectively). In a mouse Renca renal cell carcinoma model, AUNP 12 shows an additive antitumor effect when combined with Cyclophosphamide (Cat. No. 4091).
Specifications
Specifications
| Gene ID (Entrez) | 5133 |
| Purity | 95% |
| CAS | 1353563-85-5 |
| Content And Storage | Store at -20°C |
| Gene Alias | B7-DC, B7H1, B7-H1, Butyrophilin-like Protein, CD273, CD274, CD279, CD279 antigen, hPD-1, IFG, IFI, IFNG, IFNgamma, PD1, PD1hPD-l, PDCD1, PDCD1L1, PDCD1LG1, PDCD1LG2, PDL1, PDL2, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Molecular Weight (g/mol) | M.W. (Observed): 3261.56 |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

