Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AUP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-510 Supplier Invitrogen™ Supplier No. PA118257
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.

AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain they may be involved in binding ubiquitin-conjugating enzymes. The unprocessed precursor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AUP1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene AUP1
Gene Accession No. A1L134, Q9Y679
Gene Alias AA589454; ancient ubiquitous protein 1; ancient ubiquitous protein 1 isoform 2; ancient ubiquitous protein 1; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; Aup1; AUP1 lipid droplet regulating VLDL assembly factor; AUP1, lipid droplet regulating VLDL assembly factor; HUBCEP52; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; RPL40
Gene Symbols AUP1
Host Species Sheep
Immunogen Synthetic peptide corresponding to residues H(48) V F L V S C A L P D S V(60) of human ubiquitin.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101119707, 509130, 550, 680423
Target Species Human, Rat, Bovine, Sheep
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.