Description
- Dual purpose safety filter is critical to ensuring operator and laboratory safety as it provides adequate reserve gas phase capacity as the primary filter(s) reach saturation.
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected.
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR).
Specifications
Specifications
| Filter Type | Secondary |
| For Use With (Equipment) | Aura 250 Ductless Fume Hood |
| Product Type | Filter |
| Material | Gas Phase Carbon |
| For Use With (Application) | Hydrocarbons |
| Product Line | Safety Filter |