Description
- Dual purpose safety filter is critical to ensuring operator and laboratory safety as it provides adequate reserve gas phase capacity as the primary filter(s) reach saturation.
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected.
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Filter Type | Secondary |
| For Use With (Equipment) | Aura 42 and Aura 550 Fume Hood |
| Product Type | Filter |
| Material | Gas Phase Carbon |
| For Use With (Application) | Hydrocarbons |
| Product Line | Safety Filter |