Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Aura™ Series N95 Particulate Respirators

Catalog No. 19041427 Shop All 3M Products
Change view
Click to view available options
Model:
9210
9210-Plus
9211-PLUS
Size:
Small
Standard
With Exhalation Valve:
No
Yes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size With Exhalation Valve
19-041-427 9210 Small No
19-164-104 9211-PLUS Standard Yes
17-624-139 9210-Plus Standard No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-041-427 Supplier 3M™ Supplier No. 9210
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Particulate respirators with or without Cool Flow™ valve offer a comfortable fit that conform to a wide range of face sizes.

  • Available with or without Cool Flow™ one-way exhalation valve for easy exhalation and cool, dry comfort in work settings that involve heat, humidity, or long periods of wear
  • Three-panel construction resists collapse and provides better visibility
  • Soft cover web on inner panel feels soft against skin
  • Standard adjustable nose clip for custom fit and secure seal

Applications:

Grinding, bagging, sanding, sweeping and other dusty operations

Order Info

Individually packaged to prevent contamination and for easy dispensing.

TRUSTED_SUSTAINABILITY

Specifications

Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications/Compliance NIOSH TC-84A-2669
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-5668
Certifications (OSHA) OSHA 1910.134
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type N95
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size Small
With Exhalation Valve No
Filter Efficiency 95%
For Use With (Application) Grinding, sanding, sweeping, bagging and other dusty operations
Latex Free Yes
Model 9210
Nose Piece Material Aluminum
Nosepiece Type Adjustable Nose Clip
Packaging Type Individually wrapped
Quantity 20/Pk.
Sterility Non-sterile
With Faceseal No
Strap Attachment Type Stapled Braided Headbands
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.