Learn More
Selleck Chemical LLC Auranofin S4307-1g
SDP
Supplier:  Selleck Chemical LLC S43071G
Encompass_Preferred
Auranofin is an inhibitor of thioredoxin reductase (TrxR) with IC50 of 88 nM for purified H pylori TrxR in cell-free assay Auranofin has anti-cancer activity and can completely inhibit bacterial growth at 1 2 M Auranofin is an FDA-approved gold-containing compound used for the treatment of rheumatoid arthritis
Catalog No. 50-311-0267