Learn More
Abcam Auranofin, Glutathione S-transferase Inhibitor, Solid, 50 mg
Description
- Molecular Weight: 678.5 Da
- Purity: >98%
- Description: Organic gold compound, anti-inflammatory and antiproliferative agent
- Inhibitor: Glutathione S-transferase P1-1 (IC50 = 33 μM)
- Mechanism: Antiproliferative effects through STAT3 inhibition
- Form: Solid
- Activity: Orally active, shows anti-inflammatory effects in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Antioxidant |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | Glutathione S-transferase P1-1, STAT3 |
| Formulation | 100% Auranofin |
| Quantity | 50 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.