Learn More
Medchemexpress LLC Aurintricarboxylic acid ammonium salt | 569-58-4 | MFCD00040925 | 473.43 g/mol | C22H23N3O9 | 5G
SDP
Supplier:  Medchemexpress LLC HY1244935G
Encompass_Preferred
Aluminon (ATA) is the triammonium salt of aurintricarboxylic acid supplied as a research reagent. It is used primarily as a complexometric titration indicator for aluminium and has been reported to possess nuclease-inhibiting activity. The material is provided as a solid powder; handle and store according to supplier guidance and intended laboratory use.
Catalog No. 50-002-36930