Learn More
Electron Microscopy Sciences Aurion BSA-C (Acetylated BSA) 100 ML
SDP
Supplier:  Electron Microscopy Sciences 25558
Encompass_Preferred
In the blocking step, hydrophobic moieties causing "stickiness" in the specimen surface are rendered hydrophilic to minimize background. Nevertheless, the more dynamic charge-based interaction between the specimen surface and immunoreagents also needs to be controlled in order to eliminate background.
Aurion has developed BSA-c , a unique incubation buffer additive with an unparalleled ability to effectively prevent charge based background. BSA-c is prepared by acetylation of bovine serum albumin (BSA). Polycationic sites in the specimen interact readily with negatively charged acetylated BSA molecules. This significantly reduces the risk that such sites might bind negatively-charged immunoreagents and immunogold conjugates and thus reduces the risk of background.
Catalog No. 50-248-06
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.